Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD354 (TREM-1) Monoclonal Antibody (TR3MBL1), eFluor™ 660, eBioscience™
GREENER_CHOICE

Catalog No. 502090111
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-209-0111 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-209-0111 Supplier Invitrogen™ Supplier No. 50354182
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

Description: The TR3MBL1 monoclonal antibody recognizes mouse CD354 (also known as TREM-1), a 30-kDa glycosylated member of the Ig superfamily. CD354 is expressed on the cell surface of myeloid cells such as neutrophils, monocytes, dendritic cells, some macrophages, platelets, and megakaryocytes. Outside the immune system, CD354 is expressed on hepatocytes, osteoclasts, and microglia. CD354 also exists as a soluble receptor (sTREM-1) that arises from proteolytic cleavage of the membrane-bound receptor by matrix metalloproteases. CD354 plays a critical role in the innate immune response to bacterial and fungal infection by triggering the release of pro-inflammatory chemokines and cytokines. The cell surface receptor associates constitutively with the ITAM-containing adaptor protein DAP12 to activate the MAPK and NFkB signaling pathways. Finally, levels of the soluble receptor are increased in several inflammatory diseases, including sepsis, asthma, and inflammatory bowel disease, making this receptor a valuable diagnostic biomarker. Applications Reported: This TR3MBL1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This TR3MBL1 antibody has been tested by flow cytometric analysis of mouse peripheral blood cells. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but ca...

TREM1 (triggering receptor expressed on myeloid cells 1) is a receptor belonging to the Ig superfamily that amplifies neutrophil and monocyte-mediated inflammatory responses triggered by bacterial and fungal infections by stimulating release of pro-inflammatory chemokines and cytokines, as well as increased surface expression of cell activation markers. It is thought that TREM1 other related TREM proteins such as TREM2 act to modulate the inflammatory response. TREM1 has also been linked to inflammatory bowel disease resulting in enhanced production of TNF, IL-6, IL-8, and MCP-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD354 (TREM-1)
Applications Flow Cytometry
Classification Monoclonal
Clone TR3MBL1
Concentration 0.2 mg/mL
Conjugate eFluor 660
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene TREM1
Gene Accession No. Q9JKE2
Gene Alias CD354; sCD354; soluble CD354; soluble TREM 1; soluble TREM1; sTREM 1; sTREM1; TREM; Trem1; TREM-1; triggering receptor expressed in monocytes 1; triggering receptor expressed on monocytes 1; Triggering receptor expressed on myeloid cells; Triggering receptor expressed on myeloid cells 1
Gene Symbols TREM1
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58217
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Can the eFluor Organic fluorochromes be used for intracellular staining?

Yes, the eFluor Organic fluorochromes can be used for intracellular staining. The eFluor organic fluorochromes maintain bright signal and require minimal changes in compensation when fixed with eBioscience IC Fixation Buffer (Cat. No. 00-8222-49) and Permeabilization Buffer (Cat. No. 00-8333-56) or 1-step Fix/Lyse Solution (Cat. No. 00-5333-54, 00-5333-57) (as compared to live cells).

Is the eFluor 660 fluorochrome compatible with Anti-Cy5/Alexa Fluor 647 beads?

Yes, in-house studies have demonstrated that the eFluor 660 fluorochrome is recognized by Anti-Cy5/Alexa Fluor 647 beads. Side by side studies with Alexa Fluor 647 versus eFluor 660 conjugated antibodies have demonstrated comparable results.

What buffers are compatible with the eFluor Organic fluorochromes and eVolve QDots?

The eFluor Organic fluorochromes and eVolve QDots can be used with flow staining buffers containing PBS and protein.

What is the difference between eFluor Organic Dyes and eVolve Qdots?

The eFluor Organic Dyes (eFluor 450, APC-eFluor 780, PerCP-eFluor 710, eFluor 710) are conventional fluorochromes. In contrast, the eVolve line of products are Quantum dots.

Are the eFluor Organic Dyes photo-labile?

As with other fluorochromes, we recommend minimal exposure to light to maintain optimal signal.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.