Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD360 (IL-21 Receptor) Monoclonal Antibody (eBio4A9), Biotin, eBioscience™
GREENER_CHOICE

Catalog No. 5011726
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-117-26 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-117-26 Supplier Invitrogen™ Supplier No. 13121981
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

Description: The IL-21 receptor (IL-21R), also known as CD360, combines with the common gamma-chain to form a functional receptor for IL-21. IL-21R is expressed on several lineages including both resting and activated B, T, natural killer and dendritic cells. Binding of its ligand, IL-21, in these cells results in the activation of the Jak/Stat signal transduction pathway. The biological effects of ligand binding are diverse and depend on the cellular context and differentiation state of the cell. In the mouse, IL-21 binding to IL-21R induces the apoptosis of resting and activated B cells, enhances anti-CD3-mediated proliferation of T cells, and inhibits the maturation of dendritic cells and the IL-15-mediated expansion of NK cells. Applications Reported: This eBio4A9 antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBio4A9 antibody has been tested by flow cytometric analysis of mouse splenocytes, thymocytes and bone marrow. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.

The protein encoded by this gene is a cytokine receptor for interleukin 21 (IL21). It belongs to the type I cytokine receptors, and has been shown to form a heterodimeric receptor complex with the common gamma-chain, a receptor subunit also shared by the receptors for interleukin 2 (IL2) and interleukin 5 (IL5). This receptor transduces the growth promoting signal of IL21, and is important for the proliferation and differentiation of T cells, B cells, and natural killer (NK) cells. The ligand binding of this receptor leads to the activation of multiple downstream signaling molecules, including JAK1, JAK3, STAT1, and STAT3. Knockout studies of a similar gene in mouse suggest a role for this gene in regulating immunoglobulin production. Three alternatively spliced transcript variants encoding the same protein have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD360 (IL-21 Receptor)
Applications Flow Cytometry
Classification Monoclonal
Clone eBio4A9
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Il21r
Gene Accession No. Q9JHX3
Gene Alias CD360; IL-21 receptor; Il21r; IL-21R; interleukin 21 receptor; interleukin-21 receptor; LR-beta; lymphocyte receptor beta; MGC10967; NILR; novel cytokine receptor 8; Novel interleukin receptor; NR8; UNQ3121/PRO10273
Gene Symbols Il21r
Host Species Rat
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60504
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.