Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD366 (TIM-3) Mouse anti-Mouse, PE, Clone: 5D12/TIM-3 (also known as 5D12), BD

Catalog No. BDB566346 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566346 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566346 Supplier BD Biosciences Supplier No. 566346
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 5D12 monoclonal antibody specifically binds to NKp80 which is also known as C-type lectin domain family 5 member C (CLEC5C). NKp80 is a single-pass type II transmembrane glycoprotein that contains a C-type lectin extracellular domain and is expressed as a ñ80 kDa disulfide-linked homodimer. NKp80 is encoded by KLRF1 /(Killer cell lectin-like receptor subfamily F member 1) which maps to the NK-gene complex (NKC). NKp80 is expressed on most NK cells, and subsets ofγδ T cellsand effector-memory CD8αβT cells. NKp80 is not present in rodents. NKp80 is an activating coreceptor that induces NK cell-mediated cytotoxicity and proinflammatory cytokine (eg, IFN-γand TNF) production upon interaction with its ligand, AICL (Activation induced C type lectin). AICL is encoded by CLEC2B /that is selectively expressed on myeloid cells including activated monocytes, macrophages, and granulocytes. NKp80 can reportedly transduce signals through an atypical hemi-ITAM-like sequence motif and Syk kinase to trigger cellular responses.

R-phycoerythrin (PE) is an accessory photosynthetic pigment found in red algae. It exists in vitro as a 240-kDa protein with 23 phycoerythrobilin chromophores per molecule. This makes PE one of the brightest fluorochromes for flow cytometry applications, but its photobleaching properties make it unsuitable for fluorescence microscopy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD366 (TIM-3)
Classification Monoclonal
Clone 5D12/TIM-3 (also known as 5D12)
Concentration 0.2mg/mL
Conjugate PE
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Cd366, T-cell membrane protein, Tim3, TIMD-3, Timd3, HAVcr-2, Havcr2,
Host Species Mouse
Immunogen Mouse TIM-3 Recombinant Protein
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.