Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD369 (Clec7a, Dectin-1) Monoclonal Antibody (bg1fpj), PE-Cyanine7, eBioscience™
GREENER_CHOICE

Catalog No. 501123397
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-3397 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-112-3397 Supplier Invitrogen™ Supplier No. 25585980
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

The monoclonal antibody bg1fpj recognizes mouse CD369, also known as Dectin-1 or CLEC7A, a type II transmembrane protein in the C-type lectin family. The ligand is beta1, 3-glycan. Beta glycans are a main component in polysaccharides and specifically beta1,3-linked carbohydrates are found in the cell wall of fungi, plants and some bacteria. CD369 has been found to bind Candida, Aspergillis, Pneumocystis, Coccidioides and Mycobacterium. Calcium dependent binding to ligand signals through syk via the ITAM domain in the cytoplasmic portion and results in endocyotosis/phagocytosis and production of cytokines (IL-2, IL-6, IL-10, TNF alpha, IL-1 beta, IL-23). A direct role in Th17 and Th1 differentiation has been attributed to CD369 signaling. CD369 is expressed predominantly on myeloid cells (neutrophils, monocytes, macrophages, and dendritic cells). Low levels have also been reported on lymphocytes and mucosal epithelium. Applications Reported: This bg1fpj antibody has been reported for use in flow cytometric analysis. Applications Tested: This bg1fpj antibody has been tested by flow cytometric analysis of mouse lysed whole blood cells. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for opti...

CLEC7A, also known as dectin-1, is a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily and is predominantly expressed on myeloid cells. It is a small glycoprotein type II membrane receptor with an extracellular C-type lectin-like domain fold and a cytoplasmic domain with an immunoreceptor tyrosine-based activation motif (ITAM). CLEC7A functions as a pattern-recognition receptor that recognizes a variety of beta-1,3-linked and beta-1,6-linked glucans from fungi and plants, and in this way plays a role in innate immune response. Upon fungal exposure, CLEC7A activates Syk tyrosine kinase, triggering a massive oxidative burst through the formation of reactive oxygen species.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD369 (Clec7a, Dectin-1)
Applications Flow Cytometry
Classification Monoclonal
Clone bg1fpj
Concentration 0.2 mg/mL
Conjugate PE-Cyanine7
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Clec7a
Gene Accession No. Q6QLQ4
Gene Alias Beta-glucan receptor; beta-GR; BGR; CANDF4; CD369; CLEC7A; Clec7b; CLECSF12; C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 12; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 12; C-type lectin domain family 7 member A; C-type lectin domain family 7, member A; C-type lectin superfamily member 12; DC-associated C-type lectin 1; DECTIN1; dectin-1; Dendritic cell-associated C-type lectin 1; dendritic cell-associated C-type lectin-1; lectin-like receptor 1; RGD1565140; SCARE2; UNQ539/PRO1082
Gene Symbols Clec7a
Host Species Rat
Immunogen Baculovirus-expressed Fc-containing protein.
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56644
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.