Learn More
Invitrogen™ CD37 Monoclonal Antibody (IPO-24)
Description
MA1-7627 detects CD37 from human samples. MA1-7627 has been successfully used in ELISA, flow cytometry, immunoprecipitation and immunohistochemistry procedures. The MA1-7627 immunogen is a A B cell line.
Specifications
Specifications
| Antigen | CD37 |
| Applications | ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation |
| Classification | Monoclonal |
| Clone | IPO-24 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.2 |
| Gene | CD37 |
| Gene Accession No. | P11049 |
| Gene Alias | CD37; CD37 antigen; CD37 molecule; cell differentiation antigen 37; GP52-40; Leukocyte antigen CD37; leukocyte surface antigen CD37; tetraspanin-26; Tspan26; Tspan-26 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.