Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD370 (Clec9A) Monoclonal Antibody (9A11), PE, eBioscience™

Catalog No. 12370942
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12370942 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12370942 Supplier Invitrogen™ Supplier No. 12370942
  • $438.00
    $328.65 / Each of 1

    Save $109.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

This monoclonal antibody 9A11 reacts with human CD370, also known as C-type lectin family member 9A (Clec9A) and DNGR1. This type II transmembrane glycoprotein is present at the cell surface in the form of a homodimer. CD370 is a cellular receptor expressed by myeloid cells that recognizes F-actin, a dead cell-associated antigen that is exposed in necrotic cells. Upon ligation, CD370 recruits Syk kinase to the ITAM-like motif in its intracellular tail and facilitates endocytosis of antigenic material that is subsequently processed and cross-presented to CD8+ T cells. Human CD370 is expressed by CD141+ (BDCA3+) dendritic cells and, to a lesser degree, by CD14+ CD16- monocytes. Unlike in mice, human plasmacytoid dendritic cells do not express CD370. This 9A11 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Filtration: 0.2 μm post-manufacturing filtered.

C-type lectin domain family 9 member A (CLEC9A) is a gene located on chromosome 12p13.2, encoding a protein that belongs to the C-type lectin receptor family. CLEC9A is primarily expressed on dendritic cells and certain subsets of macrophages, playing a pivotal role in the immune system by recognizing and binding dead cell debris, including necrotic cells. This receptor facilitates the presentation of antigens derived from dead cells to T cells, thus contributing to the initiation of immune responses. CLEC9A is involved in cross-presentation, a process crucial for adaptive immunity, particularly in the context of viral infections and cancer. Its expression and activity are of particular interest for vaccine development and immunotherapy, given its ability to enhance antigen presentation and stimulate robust immune responses. Research into CLEC9A continues to explore its therapeutic potential for modulating immune activity in various diseases, including cancer and infectious diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD370 (Clec9A)
Applications Flow Cytometry
Classification Monoclonal
Clone 9A11
Concentration 5 μL/Test
Conjugate PE
Formulation PBS with BSA and 0.09% sodium azide
Gene CLEC9A
Gene Accession No. Q6UXN8
Gene Alias 9830005G06Rik; CD370; Clec9a; C-type lectin domain family 9 member A; C-type lectin domain family 9, member A; Dendritic cell natural killer lectin group receptor 1; DNGR1; DNGR-1; HEEE9341; RGD1562513; UNQ9341; UNQ9341/PRO34046
Gene Symbols CLEC9A
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283420
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.