Learn More
Epredia™ Lab Vision™ CD38 Ab-3, Mouse Monoclonal Antibody
Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD38 Ab-3, Mouse Monoclonal Antibody.
$566.00 - $834.00
Specifications
Antigen | CD38 Ab-3 |
---|---|
Clone | 38C03; same as SPC32 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
MS983S1
|
Epredia™
MS983S1 |
Concentrated | 500 μL |
Each for $566.00
|
|
||||
MS983S
|
Epredia™
MS983S |
Concentrated | 1 mL |
Each for $834.00
|
|
||||
MS983R7
|
Epredia™
MS983R7 |
Prediluted | 7 mL |
Each for $768.50
|
|
||||
MS983S0
|
Epredia™
MS983S0 |
Concentrated | 100 μL | N/A | N/A | N/A | |||
MS983R1
|
Epredia™
MS983R1 |
Prediluted | 1 mL | N/A | N/A | N/A | |||
Description
CD38 is a single chain type II integral transmembrane protein, which is highly expressed on thymocytes. It is also present on activated T cells and terminally differentiated B cells (plasma cells). Other reactive cells include NK cells, monocytes, macrophages, and dendritic cells. CD38 may be detected on cells from multiple myeloma, ALL (B and T) and some AML. It is, however, not found on most mature resting peripheral lymphocytes. CD38 functions as a multicatalytic ectoenzyme serving as ADP-ribosyl cyclase, cyclic ADP-ribose hydrolase and possibly NAD+ glycohydrolase or as a cell surface receptor. Antibodies to CD38 are useful in subtyping of lymphomas and leukemias, detection of plasma cells (i.e. identification of myelomas), and as a marker for activated B and T cells.Host Species: Mouse
Clone: 38C03; same as SPC32
Isotype: IgG1
Species Reactivity: Human. Others not tested.
Epitope: Extracellular
Immunogen: Recombinant protein encoding the extracellular domain of human CD38
Molecular Weight: 45 kDa
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting (Not suitable)
Note:
USA: RUO; Int'l: RUO
Specifications
CD38 Ab-3 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
IgG1 |
38C03; same as SPC32 | |
Monoclonal | |
Mouse | |
Cell Adhesion | |
Recombinant protein encoding the extracellular domain of human CD38 | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.