Learn More
Medchemexpress LLC HY-123999 5mg Medchemexpress, CD38 inhibitor 1 CAS:1700637-55-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1239995MG
Encompass_Preferred
Medchemexpress, HY-123999 5mg CD38 inhibitor 1 CAS:1700637-55-3 CD38 inhibitor 1 (compound 78c) is a potent CD38 inhibitor with IC50s of 7.3 nM and 1.9 nM for hCD38 and mouse CD38. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9656
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.