Learn More
CAPRICO BIOTECHNOLOGIES INC CD38 (MF450) Antibody
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4008145
Encompass_Preferred
The Caprico CD38 (MF450) Antibody is a mouse monoclonal antibody that recognizes human CD38. CD38 is a transmembrane glycoprotein expressed on various hematopoietic cells including T cells, B cells, NK cells, monocytes, macrophages, granulocytes, and dendritic cells. CD38 functions in cell adhesion, signal transduction and calcium signaling. This antibody is conjugated to Miltenyi Biotec's proprietary MF450 fluorochrome and is suitable for flow cytometry applications.
Catalog No. 50-227-9134
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.