Learn More
Description
MA1-7653 detects CD38 from human samples. MA1-7653 has been successfully used in ELISA and Immunoprecipitation procedures. The MA1-7653 immunogen is Hematopoietic precursors, thymocytes, activated T-cells and plasma cells. Store at 4°C short term. Store at -80°C long term.
Specifications
Specifications
| Antigen | CD38 |
| Applications | ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | ICO-20 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.2 |
| Gene | CD38 |
| Gene Accession No. | P28907 |
| Gene Alias | 2'-phospho-ADP-ribosyl cyclase; 2'-phospho-ADP-ribosyl cyclase/2'-phospho-cyclic-ADP-ribose transferase; 2'-phospho-cyclic-ADP-ribose transferase; ADPRC 1; ADPRC1; ADP-ribosyl cyclase 1; ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 1; cADPr hydrolase 1; Cd38; CD38 antigen; CD38 antigen (ADP-ribosyl cyclase / cyclic ADP-ribose hydrolase); CD38 antigen (p45); CD38 antigen p45; CD38 molecule; CD38H; Cd38-rs1; cluster of differentiation 38; cyclic ADP-ribose hydrolase 1; ecto-nicotinamide adenine dinucleotide glycohydrolase; I-19; NAD(+) nucleosidase; NAD+ nucleosidase; NIM-R5 antigen; T10 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.