Learn More
CD3e Monoclonal Antibody (M2AB), APC, Invitrogen™
Description
Specifications
Specifications
| Antigen | CD3e |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | M2AB |
| Conjugate | APC |
| Gene | CD3E |
| Gene Accession No. | P07766 |
| Gene Alias | FLJ18683; T3E; TCRE; CD3-epsilon; CD3e antigen; epsilon polypeptide TiT3 complex; T-cell antigen receptor complex; epsilon subunit of T3; T-cell surface antigen T3/Leu-4 epsilon chain; T-cell surface glycoprotein CD3 epsilon chain; HCD3 |
| Gene Symbols | CD3E |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.