Learn More
Genscript Corporation CD3ECD3G hFc Chimera Biotin
Supplier:  Genscript Corporation Z04360500
Encompass_Preferred
T-cell surface glycoprotein CD3 epsilon CD3 gamma chain also known as CD3E CD3G are single-pass type I membrane proteins When antigen presenting cells (APCs) activate T-cell receptor (TCR) TCR-mediated signals are transmitted across the cell membrane by the CD3 chains CD3D CD3E CD3G and CD3Z All CD3 chains contain immunoreceptor tyrosine-based activation motifs (ITAMs) in their cytoplasmic domain
Catalog No. 50-000-58931
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.