Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD4/CD26 Antibody Cocktail, FITC, PE
GREENER_CHOICE

Catalog No. 01670837 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 Tests
50 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-837 100 Tests
01-670-838 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-670-837 Supplier Invitrogen™ Supplier No. MA112118
Add to Cart
Add to Cart

Antibody Cocktail

This antibody is a monoclonal mixture of FITC-conjugated CD4 (clone 7E14, isotype IgG1) and PE-conjugated CD26 (clone EUG-6, isotype IgG2a).

The CD4 antigen is involved in the recognition of the type II MHC antigen. It is also a receptor for HIV. It is present on most T helper cells and normal thymocytes. The cytoplasmic portion of CD4 is associated with p56lck tyrosine kinase. CD4 expression is commonly found in human lymph nodes and tonsils. CD26 [also known as dipeptidyl peptidase IV (DPP IV), adenosine deaminase (ADA) binding protein]. 1 - 3 is an atypical serine protease belonging to the prolyl oligopeptidase family. It is expressed on lymphocyte cells and is upregulated during T-cell activation. CD26 is also expressed on activated B cells and natural killer cells and abundantly on epithelia. CD26 is implicated in a variety of biological functions including T-cell activation, cell adhesion with extracellular matrix such as fibronectin or collagens, and in HIV infection. Cross-linking of CD26 using this antibody dramatically enhances the anti CD3-induced IL - 2 production.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD4/CD26
Applications Flow Cytometry
Classification Cocktail
Clone 7E14, EUG-6, EUG-6
Conjugate FITC/PE
Formulation PBS with 0.2% carrier protein and 0.08% sodium azide
Gene CD4
Gene Accession No. P01730, P27487
Gene Alias ADABP; ADCP2; ADCP-2; Adenosine deaminase complexing protein 2; CD26; CD4; CD4 antigen (p55); CD4 antigen p55; CD4 molecule; CD4 receptor; CD4mut; dipeptidyl peptidase 4; Dipeptidyl peptidase 4 membrane form; Dipeptidyl peptidase 4 soluble form; Dipeptidyl peptidase IV; Dipeptidyl peptidase IV membrane form; Dipeptidyl peptidase IV soluble form; dipeptidylpeptidase 4; dipeptidyl-peptidase 4; dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2); DPP IV; DPP4; DPPIV; DPP-IV; T-cell activation antigen CD26; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; TP103
Gene Symbols CD4, DPP4
Host Species Mouse
Immunogen human peripheral blood T lymphocytes.
Purification Method Protein A/G
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1803, 920
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1, IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.