Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4, Mouse Anti-Human, RY610, Clone: SK3 (also known as Leu3a), BD Horizon™

Catalog No. BDB571081 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
Conjugate:
RY610
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
BDB571081 100 Tests RY610
BDB571082 25 Tests RY610
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB571081 Supplier BD Biosciences Supplier No. 571081
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD4 Monoclonal antibody specifically detects CD4 in Human (QC Testing) samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD4
Applications Flow Cytometry
Classification Monoclonal
Clone SK3 (also known as Leu3a)
Conjugate RY610
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias L3T4; Leu3a; T-cell surface antigen T4/Leu-3 ; W3/25 ; CD4 antigen (p55)
Host Species Mouse
Immunogen Human Peripheral Blood T Cells
Purification Method The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 920
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.