Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD4 Rat anti-Mouse, BUV395, Clone: GK1.5, BD Horizon™

Catalog No. BDB565974 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565974 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565974 Supplier BD Biosciences Supplier No. 565974
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

CD4 Monoclonal antibody specifically detects CD4 in Mouse samples. It is validated for Flow cytometry.
Storage Buffer: Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD4
Applications Flow Cytometry
Classification Monoclonal
Clone GK1.5
Concentration 0.2mg/mL
Conjugate Brilliant Ultraviolet 395
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias Cd4; CD4 antigen; L3T4; Ly-4; T-cell surface antigen T4/Leu-3
Host Species Rat
Immunogen Mouse CTL clone V4
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity and unconjugated antibody and free BD Horizon™ BUV395 were removed. chromatography. The antibody was conjugated with BD Horizon™ BUV395 under optimum conditions
Form Purified
Isotype IgG2b κ
Reactivity QC Testing: Mouse
Show More Show Less

For Reseach Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.