Learn More
ANCELL CORPORATION CD40 Human-Murine IgG2a Fusion Protein, Preservative Free, >95% Purity, 25 µg
Supplier:  ANCELL CORPORATION 504820
Encompass
The CD40 Human-Murine IgG2a Fusion Protein is a highly purified, preservative-free protein designed for immunological research. It facilitates studies on B cell activation and isotype switching by blocking anti-human CD40 binding and engaging with CD154 on cells. This unique fusion protein combines human CD40 with murine IgG2a Fc, offering exceptional purity (>95%). Its effective use in research applications, coupled with its stable formulation, sets it apart from other products.
Catalog No. NC2223121
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.