Learn More
Invitrogen™ CD40 Monoclonal Antibody (1C10), Brilliant Ultra Violet™ 661, eBioscience™, Invitrogen™
Description
Specifications
Specifications
| Antigen | CD40 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 1C10 |
| Concentration | 0.2 mg/mL |
| Conjugate | Brilliant Ultraviolet 661 |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | CD40 |
| Gene Accession No. | P27512 |
| Gene Alias | AI326936; B cell surface antigen CD40; B cell-associated molecule; B-cell surface antigen CD40; Bp50; CD antigen CD40; Cd40; CD40 antigen; CD40 antigen (TNF receptor superfamily member 5); CD40 antigen, TNF receptor superfamily member 5; CD40 molecule; CD40 molecule, TNF receptor superfamily member 5; CD40L receptor; CDw40; GP39; HIGM1; I79_006806; IGM; IMD3; Immunoglobulin M; ImmunoglobulinM; membrane protein CD40; MGC9013; p50; receptor for ligand CD154; sCD40; soluble CD40; T-BAM; T-cell differentiation antigen; Tnfrsf5; TRAP; tumor necrosis factor receptor superfamily member 5; tumor necrosis factor receptor superfamily, member 5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.