Learn More
Enzo Life Sciences CD40L (soluble) (human), (recombinant) (10 µg)
SDP
Supplier:  Enzo Life Sciences ALX5220152010
Encompass_Preferred
Increases the stimulation of B cells. Alternative name: CD154, TNFSF 5, gp39. MW: ~18kDa (SDS-PAGE). Concentration: 0.1mg/ml after reconstitution. Endotoxin content: E. coli. The extracellular domain of human CD40L (CD154) (aa 116-261) is fused at the N-terminus to a linker peptide (6 aa) and a FLAG-tag. Specificity: Binds to human CD40. Use/Stability: Stable for at least 6 months after receipt when stored at -20°C. UniProt: P29965. Application: ELISA. Handling: Avoid freeze/thaw cycles. After reconstitution, prepare aliquots and store at -20°C. Long Term Storage: -20°C
Catalog No. NC1597449
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.