Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD40L/CD154 Mouse anti-Human, Clone: 1E6D10, Proteintech
SDP

Catalog No. 501736925 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6925 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6925 Supplier Proteintech Group Inc Supplier No. 665021IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The CD40 antigen is a single chain glycoprotein. It is known to be a member of the tumor necrosis factor / nerve growth factor superfamily and shows a significant homology to the Hodgkin′s disease associated antigen, CD30. CD40 is present on all B cells except plasma cells. It is also found on some epithelial cells, carcinomas and lymphoid dendritic cells. The precise function of the CD40 antigen is unknown but it appears to be involved in the transduction of regulatory signals for cellular functions such as B cell proliferation and differentiation.

Specifications

Antigen CD40L/CD154
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1E6D10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CD40LG
Gene Accession No. P29965
Gene Alias CD154, CD40 L, CD40 ligand, CD40L, CD40LG, gp39, hCD40L, HIGM1, IGM, IMD3, T BAM, T cell antigen Gp39, TNF related activation protein, TNFSF5, TRAP
Gene Symbols CD40LG
Host Species Mouse
Immunogen CD40L/CD154 Fusion Protein Ag24941
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 959
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.