Learn More
AdipoGen CD40L ratmultimericrec
SDP
Supplier:  AdipoGen AG40B01073010
Encompass_Preferred
Protein. Rat CD40L aa 115-260 is fused at the N-terminus to mouse ACRP30headless aa 18-111 and a FLAG -tag. Source CHO cells. Endotoxin content <0.02EU/µg purified protein LAL test Lonza. Lyophilized. Contains PBS. Binds to rat, human and mouse CD40. Purity >95% SDS-PAGE. MultimericCD40L™ is a high activity construct in which two trimeric CD40 ligands are artificially linked via the collagen domain of ACRP30. This construct very effectively simulates the natural membrane-assisted aggregation of CD40L in vivo. It provides a simple and equally potent alternative to CD40Lenhancer combinations. MultimericCD40L™ has been shown to suppress alum-induced IL-1beta release and caspase-1 activation in a dose-, CD40- and time dependent manner, without affecting BMDM Bone marrow-derived macrophages viability. It also effectively suppressed the inflammasome function triggered by NLRP3 activators.
Catalog No. 50-256-2918
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.