Learn More
Invitrogen™ CD41 Monoclonal Antibody (HIP2)

Description
This antibody reacts with an extracellular epitope on beta (b) subunit of CD41 glycoprotein (light chain; 23 kDa). CD41 is mainly expressed on platelets and megakaryocytes.
Specifications
Specifications
| Antigen | CD41 |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | HIP2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 15mM sodium azide; pH 7.4 |
| Gene | Itga2b |
| Gene Accession No. | P08514 |
| Gene Alias | AI172977; alpha IIb; alphaIIb; alphaIIb protein; BDPLT16; BDPLT2; CD41; CD41B; form 1; form 2; GP IIb; GP2B; GPalpha IIb; GPIIb; GT; GTA; HPA3; integrin alpha 2b; integrin alpha 2b (Cd41b); integrin alpha-IIb; Integrin alpha-IIb heavy chain; Integrin alpha-IIb light chain; Integrin alpha-IIb light chain, form 1; Integrin alpha-IIb light chain, form 2; integrin subunit alpha 2b; integrin subunit alpha IIb; integrin, alpha 2B; integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41); ITGA2B; ITGAB; platelet fibrinogen receptor, alpha subunit; platelet glycoprotein IIb; platelet glycoprotein IIb of IIb/II Ia complex; platelet glycoprotein IIb of IIb/IIIa complex; platelet membrane glycoprotein IIb; platelet-specific antigen BAK; PPP1R93; protein phosphatase 1, regulatory subunit 93 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.