Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Probes™ CD44 Alexa Fluor™ 488 Conjugate Kit for Live Cell Imaging

Catalog No. A25528
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25528 50 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A25528 Supplier Molecular Probes™ Supplier No. A25528

CD44 Alexa Fluor™ 488 Conjugate Kit for Live Cell Imaging

Molecular Probes™ human pluripotent stem cell (PSC) live imaging kits provide reagents for superior imaging of live human PSCs. This CD44 Alexa Fluor™ 488 Conjugate Kit includes an optimally labeled primary antibody paired with reduced-background imaging medium (FluoroBrite™ DMEM) for the specific detection of the negative PSC marker CD44. For fixed cell staining of PSC markers and germ layer markers, we recommend the Pluripotent Stem Cell 4-Marker Immunocytochemistry Kit and the 3-Germ Layer Immunocytochemistry Kit.

With the CD44 Alexa Fluor™ 488 Conjugate Kit for Live Cell Imaging, you can:
  • Detect the expression of a negative pluripotent stem cell marker using an optimally dye-conjugated antibody
  • Preserve cell health and maximize fluorescence signal with FluoroBrite™ DMEM imaging medium
  • Stain live cell samples with confidence using reagents that are tested to be free of common contaminants
  • Combine with additional markers to get more information per sample

    Detect Expression of a Negative PSC Marker
    Powered by the high performance of bright, photostable Alexa Fluor™ dyes, this kit enables reliable live-cell detection of CD44. CD44 is absent in PSCs and expressed on the surface of many other cell types, including human and mouse fibroblasts. This antibody conjugate has been demonstrated to show superior staining results when used with FluoroBrite™ DMEM.

    Preserve Cell Health and Maximize Signal
    Staining cells in FluoroBrite™ DMEM maximizes cell health and enhances signal-to-noise ratio when performing live-cell fluorescence imaging.

    Stain Live-cell Samples with Confidence
    Unlike most other conjugated antibodies, the reagents provided in this kit are 0.2μm sterile-filtered and tested to be free of common contaminants.

    Combine with Other Markers
    To get more information per sample, combine this kit with the TRA-1-60 Alexa Fluor™ 555 Conjugate Kit for Live Cell Imaging or the TRA-1-60 Alexa Fluor™ 594 Conjugate Kit for Live Cell Imaging.

    What You Get
    Easily scaled to any culture plate, dish, or slide format, the CD44 Alexa Fluor™ 488 Conjugate Kit comes with reagents sufficient to stain 50 samples using a 200μL staining volume and includes:
  • 200μL of Alexa Fluor™ 488 Rat Anti-Human and Mouse CD44 (50X concentration)
  • 500 mL FluoroBrite™ DMEM
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage
    • 200 μL of Alexa Fluor™ 488 Rat Anti-Human and Mouse CD44 (50x concentration) (store at 2°C to 8°C)
    • 500 mL FluoroBrite™ DMEM
    • Store at 2°C to 8°C, protect from light
    Dye Type Alexa Fluor™ 488
    Quantity 50 tests
    Product Line Alexa Fluor
    Product Type Conjugate Kit

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.