Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CD44 Pre-design Chimera RNAi

Catalog No. 89035711 Shop All Abnova Corporation Products
Change view
Click to view available options
Description:
Homo sapiens CD44 antigen (homing function and Indian blood group system) (CD44), transcript variant 8, mRNA.
Homo sapiens CD44 antigen (homing function and Indian blood group system) (CD44), transcript variants, mRNA.
Quantity:
10 nmol
20 nmol
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Quantity
89-035-711 Homo sapiens CD44 antigen (homing function and Indian blood group system) (CD44), transcript variants, mRNA. 20 nmol
89-035-708 Homo sapiens CD44 antigen (homing function and Indian blood group system) (CD44), transcript variant 8, mRNA. 10 nmol
89-035-709 Homo sapiens CD44 antigen (homing function and Indian blood group system) (CD44), transcript variant 8, mRNA. 20 nmol
89-035-710 Homo sapiens CD44 antigen (homing function and Indian blood group system) (CD44), transcript variants, mRNA. 10 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 89-035-711 Supplier Abnova™ Supplier No. H00000960R12L

Homo sapiens CD44 antigen (homing function and Indian blood group system) (CD44), transcript variant 8, mRNA.

  • Gene name: CD44
  • Gene Alias: CDW44,CSPG8,ECMR-III,HCELL,IN,LHR,MC56,MDU2,MDU3,MGC10468,MIC4,MUTCH-I,Pgp1
  • Entrez GeneID: 960
  • strong>Gene Description: CD44 molecule (

Specifications

Content And Storage Store at -20°C, do not exceed 4 - 5 freeze-thaw cycles to ensure product integrity.
Description Homo sapiens CD44 antigen (homing function and Indian blood group system) (CD44), transcript variants, mRNA.
For Use With (Application) RNAi Knockdown
Includes DEPC water
Product Type CD44 Pre-design Chimera RNAi
Quantity 20 nmol
Target Human

For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.