Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ CD44 Pre-design Chimera RNAi
Homo sapiens CD44 antigen (homing function and Indian blood group system) (CD44), transcript variant 8, mRNA.
Specifications
Content And Storage | Store at -20°C, do not exceed 4 - 5 freeze-thaw cycles to ensure product integrity. |
---|---|
For Use With (Application) | RNAi Knockdown |
Includes | DEPC water |
Product Type | CD44 Pre-design Chimera RNAi |
Target | Human |
Catalog Number | Mfr. No. | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Quantity | Price | Quantity & Availability | ||||
89-035-708
|
Abnova™
H00000960R11S |
Homo sapiens CD44 antigen (homing function and Indian blood group system) (CD44), transcript variant 8, mRNA. | 10 nmol | N/A | N/A | N/A | |||
89-035-709
|
Abnova™
H00000960R11L |
Homo sapiens CD44 antigen (homing function and Indian blood group system) (CD44), transcript variant 8, mRNA. | 20 nmol | N/A | N/A | N/A | |||
89-035-710
|
Abnova™
H00000960R12S |
Homo sapiens CD44 antigen (homing function and Indian blood group system) (CD44), transcript variants, mRNA. | 10 nmol | N/A | N/A | N/A | |||
89-035-711
|
Abnova™
H00000960R12L |
Homo sapiens CD44 antigen (homing function and Indian blood group system) (CD44), transcript variants, mRNA. | 20 nmol | N/A | N/A | N/A | |||
Description
- Gene name: CD44
- Gene Alias: CDW44,CSPG8,ECMR-III,HCELL,IN,LHR,MC56,MDU2,MDU3,MGC10468,MIC4,MUTCH-I,Pgp1
- Entrez GeneID: 960
- Gene Description: CD44 molecule
Specifications
Store at -20°C, do not exceed 4 - 5 freeze-thaw cycles to ensure product integrity. | |
DEPC water | |
Human |
RNAi Knockdown | |
CD44 Pre-design Chimera RNAi |
For research use only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title