Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45.1 Mouse anti-Mouse, APC, Clone: A20, BD

Catalog No. BDB561873 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561873 25 μg
BDB558701 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB561873 Supplier BD Biosciences Supplier No. 561873
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The A20 monoclonal antibody specifically binds to CD45 (Leukocyte Common Antigen) on all leukocytes of mouse strains expressing the CD45.1 alloantigen (eg, RIII, SJL/J, STS/A, DA). This alloantigen was originally named Ly-5.2, and this was the designation at the time that the antibody was characterized. The designation was later changed from Ly-5.2 to Ly-5.1 to conform with the convention that the .2 alloantigen designations be assigned to the C57BL/6 strain. mAb A20 has been reported not to react with leukocytes of most other mouse strains, which express the CD45.2 alloantigen. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family: Its intracellular (COOH-terminal) region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated A, B, and C,respectively), plus differing levels of glycosylation. The CD45 isoforms detected in the mouse are cell type-, maturation-, and activation state-specific. The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction. The A20 antibody has been reported to inhibit some responses of B cells, from mice expressing the CD45.1 alloantigen, to antigens and LPS.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD45.1
Applications Flow Cytometry
Classification Monoclonal
Clone A20
Concentration 0.2mg/mL
Conjugate APC
Description Ly-5; Lyt4; CD45R; LCA; Ptprc; Protein tyrosine phosphatase receptor type C
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen SJL mouse thymocytes and splenocytes
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.