Learn More
Invitrogen™ CD45.2 Monoclonal Antibody (104), PE-Cyanine7

Description
CD45.2 Monoclonal Antibody for Flow
Specifications
Specifications
| Antigen | CD45.2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 104 |
| Concentration | 0.1 mg/mL |
| Conjugate | PE-Cyanine7 |
| Formulation | PBS with proprietary stabilizer and <0.1% sodium azide; pH 7.1 |
| Gene | Ptprc |
| Gene Accession No. | P06800 |
| Gene Alias | B220; Cd45; CD45 antigen; CD45R; GP180; LCA; L-CA; leukocyte common antigen; loc; LY5; Ly-5; lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; protein tyrosine phosphatase, receptor type, C; Ptprc; Receptor-type tyrosine-protein phosphatase C; T200; T200 glycoprotein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.