Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45 Mouse anti-Human, APC-Cy7, Clone: 2D1, BD

Catalog No. BDB557833 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557833 100 Tests
BDB561863 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB557833 Supplier BD Biosciences Supplier No. 557833
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

The 2D1 monoclonal antibody recognizes an epitope on all forms of CD45, a tyrosine phosphatase. CD45 is a type I transmembrane glycoprotein that belongs to the Protein Tyrosine Phosphatase (PTP) family. There are several isoforms of CD45 with molecular weights ranging from 180 to 220 kDa. CD45 is expressed on all nucleated cells of hematopoietic origin and is also known as the Leukocyte Common Antigen (LCA). CD45 is most highly expressed on lymphocytes and multiple CD45 isoforms can be coexpressed on individual leucocytes. CD45 plays an important role in regulating leucocyte functions by modifying other signaling molecules.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD45
Applications Flow Cytometry
Classification Monoclonal
Clone 2D1
Conjugate APC-Cyanine7
Description PTPRC; CD45R; LCA; L-CA; Leukocyte Common Antigen; B220; T200; GP180; LY5
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.