Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45 Rat anti-Mouse , PE, Clone: I3/2.3 (also known as I3/2 or I3), BD Biosciences™

Catalog No. BDB567111 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB567111 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB567111 Supplier BD Biosciences Supplier No. 567111
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

CD45 Monoclonal antibody specifically detects CD45 in Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD45
Applications Flow Cytometry
Classification Monoclonal
Clone I3/2.3 (also known as I3/2 or I3)
Conjugate PE
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Cd45; L-CA; LCA; Ly-5; Ly5; Ptprc
Host Species Rat
Immunogen Mouse Lymphoma Cell Line
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
Isotype IgG2b λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.