Learn More
STEMCELL Technologies Anti-Human CD45RA, Clone HI100, Biotin, Size: 100 ug
SDP
Supplier:  STEMCELL Technologies 1000322
Encompass_Preferred
The HI100 antibody reacts with an extracellular epitope on the CD45RA isoform of CD45 a type I transmembrane glycoprotein and member of the protein tyrosine phosphatase family (receptor class 1/6 subfamily). Alternative splicing of exons 4 5 and 6 that encode the extracellular RA RB and RC polypeptides of CD45 gives rise to several isoforms with molecular masses of 180 - 240 kDa. Excision of exon 4 generates the ~220-kDa CD45RA isoform which is expressed on naïve T cells subsets of B cells and monocytes and medullary thymocytes. CD45RA functions to enhance signaling through both the T cell receptor and B cell receptor. The HI100 antibody is commonly used in combination with antibodies against the CD45RO isoform to discern naïve and memory T cells and is useful for identifying the suppressor/inducer subset of CD4 lymphocytes. The proportion of CD45RA T cells typically decreases with age.
Catalog No. 50-239-0147
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.