Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45RB Monoclonal Antibody (2B11/PD7-26), Zeta

Catalog No. 502215884
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-221-5884 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-221-5884 Supplier ZETA CORPORATION Supplier No. Z2093MS
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody Cocktail

A recommended positive control tissue for this product is Lymph Node, however positive controls are not limited to this tissue type. The primary antibody is intended for laboratory professional use in the detection of the corresponding protein in formalin-fixed, paraffin-embedded tissue stained in manual qualitative immunohistochemistry (IHC) testing. This antibody is intended to be used after the primary diagnosis of tumor has been made by conventional histopathology using non-immunological histochemical stains.

CD45RB Monoclonal antibody specifically detects CD45RB in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen CD45RB
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2B11/PD7-26
Conjugate Unconjugated
Formulation tris with BSA, NP-40 and <0.1% sodium azide
Gene Ptprc
Gene Accession No. P08575
Gene Alias B220; CD45; CD45 antigen; CD45R; GP180; LCA; L-CA; Leukocyte common antigen; loc; LY5; Ly-5; Lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; membrane tyrosine phosphatase; protein tyrosine phosphatase receptor type C; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; PTPRC; Receptor-type tyrosine-protein phosphatase C; T200; T200 glycoprotein; T200 leukocyte common antigen
Host Species Mouse
Immunogen PD7/26/16: human peripheral blood lymphocytes maintained in T cell growth factor; 2B11: isolated neoplastic cells from T cell lymphoma
Purification Method Protein A
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5788
Target Species Human
Content And Storage 2-8°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.