Learn More
Invitrogen™ CD45RB Monoclonal Antibody (C363.16A), PE, eBioscience™, Invitrogen™

Description
Description: The C363.16A monoclonal antibody reacts with the exon B-dependent isoform of mouse CD45. This CD45 isoform is expressed by thymocytes, peripheral B cells and a subpopulation of T cells at high density and by dendritic cells and macrophages at low density. CD45 is a protein tyrosine phosphatase and serves an important role in leukocyte signal transduction. Applications Reported: The C363.16A antibody has been reported for use in flow cytometric analysis. Applications Tested: The C363.16A antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Specifications
Specifications
| Antigen | CD45RB |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | C363.16A |
| Concentration | 0.2 mg/mL |
| Conjugate | PE |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | Ptprc |
| Gene Accession No. | P06800 |
| Gene Alias | B220; CD45; CD45 antigen; CD45R; GP180; LCA; L-CA; Leukocyte common antigen; loc; LY5; Ly-5; Lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; membrane tyrosine phosphatase; protein tyrosine phosphatase receptor type C; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; PTPRC; Receptor-type tyrosine-protein phosphatase C; T200; T200 glycoprotein; T200 leukocyte common antigen |
| Show More |
Safety and Handling
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.