Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD46 (Cyt1) Mouse anti-Human, Clone: 2F1, MilliporeSigma™

Catalog No. p-7222148 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF268325 25 μg
MABF2683100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF268325 Supplier MilliporeSigma Supplier No. MABF268325UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD46 (Cyt1) Monoclonal antibody specifically detects CD46 (Cyt1) in Human samples. It is validated for ELISA, Flow Cytometry, Immunofluorescence, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD46 (Cyt1)
Applications ELISA, Flow Cytometry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 2F1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to 28 amino acids from the C-terminal region of human Membrane cofactor protein (CD46).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for 1 year at +2°C to +8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.