Learn More
Genscript Corporation CD14 His Human
Supplier:  Genscript Corporation Z04155100
Encompass_Preferred
Human monocyte differentiation antigen CD14 is a pattern recognition receptor (PRR) that enhances innate immune responses CD14 was first identified as a marker of monocytes to signal intracellular responses upon bacterial encounters Given the absence of an intracellular tail CD14 was doubted to have the signaling capacities
Catalog No. 50-000-59004
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.