Learn More
Genscript Corporation CD47 His Cynomolgus/Rhesus m
Supplier:  Genscript Corporation Z04143100
Encompass
CD47 (Cluster of Differentiation 47) also known as integrin associated protein (IAP) is a transmembrane protein that in humans is encoded by the CD47 gene CD47 belongs to the immunoglobulin superfamily and partners with membrane integrins and also binds the ligands thrombospondin-1 (TSP-1) and signal-regulatory protein alpha (SIRP ) CD-47 acts as a don t eat me signal to macrophages of the immune system which has made it a potential therapeutic target in some cancers
Catalog No. 50-000-59015
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.