Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
specifically detects in Mouse samples. It is validated for ELISA, Sandwich ELISA.
Specifications
Specifications
| Assay Range | 46.875 pg/mL |
| Assay Sensitivity | 46.875-3000 pg/mL |
| Assay | Sandwich ELISA |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | Antibody Pairs |
| Specificity | This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse CD48/SLAMF2. |
| Target Species | Mouse |
| For Use With (Application) | Sandwich ELISA |
| Gene Alias | BCM1 surface antigen, BCM1Leukocyte antigen MEM-102, BLAST, BLAST1TCT.1, B-lymphocyte activation marker BLAST-1, CD48 antigen, CD48 antigen (B-cell membrane protein), CD48 molecule, hCD48, mCD48, MEM-102, SLAMF2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
