Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD49b (Integrin alpha 2) Monoclonal Antibody (DX5), Biotin, eBioscience™, Invitrogen™

Catalog No. 5012077
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-120-77 500 μg
50-120-76 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-120-77 Supplier Invitrogen™ Supplier No. 13597185
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Description: The DX5 monoclonal antibody reacts with CD49b, an antigen expressed on a majority of mouse natural killer cells and a subset of T cells. DX5 reacts with all strains of mouse tested including the most commonly used strains, BALB/c, C57BL/6, C3H, CBA, DBA, AKR, SJL and 129. Simultaneous staining of C57BL/6 spleen cells with anti-NK1.1 mAb (PK136) and DX5 reveals coexpression of both markers by a majority of cells as well as presence of small populations of DX5+PK136- and DX5-PK136+ cells. Applications Reported: The DX5 antibody has been reported for use in flow cytometric analysis. Applications Tested: The DX5 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.

CD49b (Integrin alpha 2, Integrin alpha 2 chain, VLA-2 alpha chain, HM alpha 2) is a member of the integrin family. It is a glycoprotein with molecular weight of 150 kD, and it complexes with CD29 (Integrin beta 1) to form the heterodimeric integrin VLA-2 (integrin alpha 2 beta 1, or GPIa/IIa) complex. VLA-2 is an extracellular receptor for laminin, collagen, and fibronectin, and interaction with its ligands results in the activation of intracellular signaling pathways. It has reported roles in VEGF-induced angiogenesis in vivo, as well as adhesion and lymphocyte activation. CD49b is expressed by NK cells, NK-T cells, monocytes, platelets, and epithelial cells. It is also expressed on adaptive immune cells such as T and B cells, specifically on a subset of CD4+ T cells in the spleen, on intraepithelial and lamina propria lymphocytes in the intestine, as well as on a population of peripheral CD4+ type 1 T regulatory (Tr1) cells that co-express LAG-3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD49b (Integrin alpha 2)
Applications Flow Cytometry
Classification Monoclonal
Clone DX5
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Itga2
Gene Accession No. Q62469
Gene Alias alpha 2 subunit of VLA-2 receptor; BDPLT9; BR; CD49 antigen-like family member B; CD49B; Collagen receptor; DX5; GPIa; HPA-5; human platelet alloantigen system 5; integrin alpha 2; integrin alpha-2; integrin subunit alpha 2; integrin, alpha 2; integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor); Itga2; platelet antigen Br; platelet glycoprotein GPIa; platelet membrane glycoprotein Ia; very late activation protein 2 receptor, alpha-2 subunit; VLA-2; VLA2 receptor; VLA-2 receptor, alpha 2 subunit; VLA-2 subunit alpha; VLAA2
Gene Symbols Itga2
Host Species Rat
Purification Method Affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16398
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgM κ
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.