Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD49b (Integrin alpha 2) Monoclonal Antibody (eBioY418 (Y418)), FITC, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 509614
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-961-4 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-961-4 Supplier Invitrogen™ Supplier No. 11049842
  • $284.00
    $213.65 / Each of 1

    Save $70.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: The eBioY418 (Y418) monoclonal antibody reacts with human CD49b, the 150-kDa, integrin alpha 2 subunit. The complex of CD49b non-covalently associated with integrin beta 1 (CD29), also known as VLA-2, is a receptor for collagen and laminin. This complex is expressed by lymphocytes, activated T cells, monocytes and platelets. Applications Reported: This eBioY418 (Y418) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBioY418 (Y418) antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

CD49b (Integrin alpha 2, Integrin alpha 2 chain, VLA-2 alpha chain, HM alpha 2) is a member of the integrin family. It is a glycoprotein with molecular weight of 150 kD, and it complexes with CD29 (Integrin beta 1) to form the heterodimeric integrin VLA-2 (integrin alpha 2 beta 1, or GPIa/IIa) complex. VLA-2 is an extracellular receptor for laminin, collagen, and fibronectin, and interaction with its ligands results in the activation of intracellular signaling pathways. It has reported roles in VEGF-induced angiogenesis in vivo, as well as adhesion and lymphocyte activation. CD49b is expressed by NK cells, NK-T cells, monocytes, platelets, and epithelial cells. It is also expressed on adaptive immune cells such as T and B cells, specifically on a subset of CD4+ T cells in the spleen, on intraepithelial and lamina propria lymphocytes in the intestine, as well as on a population of peripheral CD4+ type 1 T regulatory (Tr1) cells that co-express LAG-3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD49b (Integrin alpha 2)
Applications Flow Cytometry
Classification Monoclonal
Clone eBioY418 (Y418)
Concentration 5 μL/Test
Conjugate FITC
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene Itga2
Gene Accession No. P17301
Gene Alias alpha 2 subunit of VLA-2 receptor; BDPLT9; BR; CD49 antigen-like family member B; CD49B; Collagen receptor; DX5; GPIa; HPA-5; human platelet alloantigen system 5; integrin alpha 2; integrin alpha-2; integrin subunit alpha 2; integrin, alpha 2; integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor); Itga2; platelet antigen Br; platelet glycoprotein GPIa; platelet membrane glycoprotein Ia; very late activation protein 2 receptor, alpha-2 subunit; VLA-2; VLA2 receptor; VLA-2 receptor, alpha 2 subunit; VLA-2 subunit alpha; VLAA2
Gene Symbols Itga2
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3673
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.