Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD49c (Integrin alpha 3) Monoclonal Antibody (17C6), FITC
GREENER_CHOICE

Catalog No. PIMA528110
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528110 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528110 Supplier Invitrogen™ Supplier No. MA528110
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes a 150 kDa cell surface antigen which is also known as the alpha-3 integrin and as VLA-3. CD49c is very weakly expressed on peripheral blood leucocytes and is not found on platelets. Epithelial cells and endothelial cells express significant levels of CD49c.

ITGA3 (INTEGRIN, ALPHA-3), also called CD49C, VLA3 or GAPB3, is a protein that in humans is encoded by the ITGA3 gene. ITGA3 is an integrin alpha subunit which is also a member of the family of cell surface adhesion molecules. ITGA3 is mapped to chromosome 17 and its exact cytogenetic location is 17q21.33. ITGA3 makes up half of the alpha3beta1 integrin duplex that plays a role in neural migration and corticogenesis together with beta-1 subunit. A functional link between DAB1 phosphorylation and ITGA3 signaling drives the timely detachment of migrating neurons from radial glial fibers. Expression of human ITGA3 increased the infectivity of virus for Chinese hamster ovary cells. ITGA3 also contains 13 potential N-glycosylation sites, 2 potential cleavage sites, and the 7 N-terminal repeating units characteristic of ITGAs. Recombinant ITGA3 is expressed as a 150-kD protein as the same size as the native protein by the western blot analysis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD49c (Integrin alpha 3)
Applications Flow Cytometry
Classification Monoclonal
Clone 17C6
Concentration 0.1 mg/mL
Conjugate FITC
Formulation PBS with 1% BSA and 0.09% sodium azide
Gene ITGA3
Gene Accession No. P26006
Gene Alias AA407068; alpha 3 subunit of VLA-3 receptor; Alpha3 integrin; antigen CD49C; antigen identified by monoclonal antibody J143; CD49 antigen-like family member C; CD49c; CD49c antigen; FLJ34631; FLJ34704; FRP-2; Galactoprotein B3; GAPB3; GAP-B3; ILNEB; integrin alpha 3; Integrin alpha3; Integrin alpha-3; Integrin alpha-3 heavy chain; Integrin alpha-3 light chain; integrin subunit alpha 3; integrin, alpha 3; integrin, alpha 3 (antigen CD49C, alpha 3 subunit of VLA-3 receptor); ITGA3; MSK18; testicular tissue protein Li 96; testicular tissue protein Li 98; VCA-2; very late activation protein 3 receptor, alpha-3 subunit; VL3A; VLA-3 subunit alpha; VLA3a
Gene Symbols ITGA3
Host Species Mouse
Immunogen CD49c transfected cell line.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3675
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.