Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD49e (Integrin alpha 5) Monoclonal Antibody (eBioHMa5-1 (HMa5-1)), eBioscience™, Invitrogen™

Catalog No. 5012630
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-126-30 500 μg
50-126-29 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-126-30 Supplier Invitrogen™ Supplier No. 14049385
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Armenian Hamster Monoclonal Antibody

Description: The eBioHMalpha5-1 monoclonal antibody reacts with mouse and rat CD49e, also known as integrin alpha5 or VLA-5A. CD49e is a member of the integrin alpha chain family, and binds with CD29 (integrin beta1) to form the receptor VLA-5, which binds the ligands fibronectin and fibrinogen. CD49e is a type I, single-pass membrane protein, with a molecular weight of 135 kDa, and is expressed on multiple cell types including thymocytes, mast cells, activated T cells and splenic B cells. In addition to its role in adhesion, VLA-5 contributes to T-cell co-stimulation. The eBioHMalpha5-1 monoclonal antibody has been demonstrated to interfere with VLA-5-mediated cell adhesion, and with the degranulation of IgE-sensitized rat RBL cells stimulated on fibronectin-coated plates. Furthermore, HMalpha5-1 injected into mice subcutaneously, along with anti-CD49d and anti-CD61, is able to inhibit passive cutaneous anaphylaxis. Applications Reported: This eBioHMa5-1 (HMa5-1) antibody has been reported for use in flow cytometric analysis, and immunoprecipitation. (Please use Functional Grade purified eBioHMa5-1 (HMa5-1), cat. 16-0493, in functional assays.). Applications Tested: This eBioHMa5-1 (HMa5-1) antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL.

The product of this gene belongs to the integrin alpha chain family. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This gene encodes the integrin alpha 5 chain. Alpha chain 5 undergoes post-translational cleavage in the extracellular domain to yield disulfide-linked light and heavy chains that join with beta 1 to form a fibronectin receptor. In addition to adhesion, integrins are known to participate in cell-surface mediated signalling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD49e (Integrin alpha 5)
Applications Flow Cytometry, Functional Assay, Immunoprecipitation
Classification Monoclonal
Clone eBioHMa5-1 (HMa5-1)
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Itga5
Gene Accession No. P11688
Gene Alias 2-phospho-D-glycerate hydro-lyase; alpha enolase like 1; Alpha-enolase; alpha-enolase; c-myc promoter-binding protein-1; CD49 antigen-like family member E; Cd49e; C-myc promoter-binding protein; ENO1; ENO1L1; Enolase 1; enolase 1, (alpha); enolase-alpha; epididymis secretory protein Li 17; fibronectin receptor alpha polypeptide; fibronectin receptor subunit alpha; fibronectin receptor, alpha polypeptide; fibronectin receptor, alpha subunit; FNRA; HEL-S-17; integrin alpha 5; integrin alpha 5 (fibronectin receptor alpha); integrin alpha-5; Integrin alpha-5 heavy chain; Integrin alpha-5 light chain; Integrin alpha-F; integrin subunit alpha 5; integrin, alpha 5 (fibronectin receptor, alpha polypeptide); ITGA5; Itga5_retired; MBP-1; MBPB1; MPB1; MPB-1; MYC promoter-binding protein 1; NNE; Non-neural enolase; phosphopyruvate hydratase; Plasminogen-binding protein; PPH; tau-crystallin; very late activation protein 5, alpha subunit; VLA5; VLA-5; VLA5A
Gene Symbols Itga5
Host Species Armenian Hamster
Purification Method Affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16402, 315346
Target Species Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.