Learn More
Diagnostic Biosystems CD5 CD5 (SP19)
Supplier:  Diagnostic Biosystems RMAB01101
Encompass_Preferred
This antibody recognizes CD5, which is a transmembrane protein, is found on 95%of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. CD5 is expressed by many T cell leukemia, lymphomas, and activated T cells. Occasionally, CD5 antigen is also expressed on a subset of B cells. Mantle cell lymphomas(same as diffuse centrocytic lymphomas) are CD5(+) while the follicle center cell lymphoma are CD5(-).
Catalog No. 50-238-5358
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.