Learn More
Invitrogen™ CD5/CD20 Antibody Cocktail, FITC, PE

Description
This antibody is a monoclonal mixture of FITC-conjugated CD5 (clone M28623, isotype IgG2a) and PE-conjugated CD20 (clone DFA-7, isotype IgG1).
Specifications
Specifications
| Antigen | CD5/CD20 |
| Applications | Flow Cytometry |
| Classification | Cocktail |
| Clone | M28623, DFA-7, DFA-7 |
| Conjugate | FITC/PE |
| Formulation | PBS with 0.2% carrier protein and 0.08% sodium azide |
| Gene | MS4A1 |
| Gene Accession No. | P06127, P11836 |
| Gene Alias | B1; B-lymphocyte antigen CD20; B-lymphocyte cell-surface antigen B1; B-lymphocyte surface antigen B1; Bp35; CD20; CD20 antigen; CD20 receptor; CD5; CD5 antigen (p56-62); CD5 antigen p56-62; CD5 molecule; CVID5; LEU1; LEU-16; Leukocyte surface antigen Leu-16; lymphocyte antigen T1/Leu-1; membrane spanning 4-domains A1; membrane-spanning 4-domains subfamily A member 1; membrane-spanning 4-domains, subfamily A, member 1; MGC3969; MS4A1; MS4A2; S7; T1; T-cell surface glycoprotein CD5 |
| Gene Symbols | CD5, MS4A1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.