Learn More
CAPRICO BIOTECHNOLOGIES INC Anti-CD52 FITC Antibody
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4085015
Encompass_Preferred
The CapricoBio Anti-CD52 FITC Antibody is a fluorescein isothiocyanate (FITC) conjugated monoclonal antibody that recognizes human CD52. CD52 is a small glycoprotein expressed on the surface of mature lymphocytes, monocytes, eosinophils and epithelial cells. This antibody can be used in flow cytometry, immunofluorescence microscopy and other assays to identify and analyze CD52 expressing cells through its FITC fluorescent label. It is supplied as 100 tests in PBS with 0.1% sodium azide and 0.2% BSA. The concentration is 0.5 mg/mL.
Catalog No. 50-227-9201
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.