Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD52 Monoclonal Antibody (CF1D12), Super Bright™ 600, eBioscience™, Invitrogen™

Catalog No. 63052941
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-052-941 25 Tests
63-052-942 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 63-052-941 Supplier Invitrogen™ Supplier No. 63052941
  • $192.00
    $143.65 / Each of 1

    Save $48.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

This CF1D12 monoclonal antibody reacts with human CD52, a cell surface glycoprotein expressed in the immune and reproductive systems. CF1D12 is thought to recognize the carbohydrate region of CD52, not the protein core of the molecule. Tissue-specific mod

CD52, also known as CAMPATH-1 antigen, is a glycoprotein that is expressed on the surface of various immune cells, including T and B lymphocytes, natural killer cells, and monocytes. It is involved in the regulation of immune responses and is being studied as a potential therapeutic target in various diseases, including multiple sclerosis, chronic lymphocytic leukemia, and autoimmune disorders. CD52 is the target of alemtuzumab, a monoclonal antibody that is used to treat these conditions by depleting immune cells that express CD52.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD52
Applications Flow Cytometry
Classification Monoclonal
Clone CF1D12
Concentration 5 μL/Test
Conjugate Super Bright 600
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene CD52
Gene Accession No. P31358
Gene Alias AI463198; B7; B7-Ag; cambridge pathology 1 antigen; CAMPATH-1; CAMPATH-1 antigen; Cd52; CD52 antigen; CD52 antigen (CAMPATH-1 antigen); CD52 molecule; CDW52; CDW52 antigen (CAMPATH-1 antigen); CLS1; Epididymal secretory protein E5; epididymis secretory sperm binding protein Li 171mP; he5; HEL-S-171mP; human epididymis-specific protein 5; Lymphocyte differentiation antigen B7; Mb7
Gene Symbols CD52
Host Species Mouse
Immunogen Human CD52.
Purification Method Affinity chromatography
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1043
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG3 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.