Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD53 Recombinant Rabbit Monoclonal Antibody (116)
GREENER_CHOICE

Catalog No. PIMA528976
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528976 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528976 Supplier Invitrogen™ Supplier No. MA528976
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Cynomolgus CD53.

CD53 is a tetraspanin family transmembrane glycoprotein expressed in the lymphoid-myeloid lineage. This molecule has been reported to form complexes with other leukocyte surface proteins such as CD2, CD19, CD21, MHC II, VLA-4 or tetraspanins CD37, CD81 and CD82, thus probably modulating various signaling processes. CD53 is involved in radioresistancy of tumor cells and its triggering has anti-apoptotic effect. In thymus, CD53 is up-regulated in response to positive selection signals during T cell development, and is strongly expressed upon macrophage exposure to bacterial lipopolysaccharide, whereas stimulation of neutrophils results in down-regulation of CD53 expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD53
Applications ELISA
Classification Recombinant Monoclonal
Clone 116
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Cd53
Gene Accession No. 0
Gene Alias AI323659; antigen MOX44 identified by monoclonal antibody MRC-OX44; Cd53; CD53 antigen; CD53 glycoprotein; CD53 molecule; CD53 tetraspan antigen; cell surface antigen; cell surface glycoprotein CD53; Leukocyte antigen MRC OX-44; leukocyte surface antigen CD53; MOX44; OX44; Ox-44; tetraspanin-25; transmembrane glycoprotein; TSPAN25; tspan-25
Gene Symbols Cd53
Host Species Rabbit
Immunogen Recombinant Cynomolgus CD53 Protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102127943
Target Species Cynomolgus Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.