Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Horizon™ CD54 (ICAM-1) Mouse, anti-Human, RY586, Clone: HA58, BD Horizon™

Catalog No. p-7225538 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB568122 25 Tests
BDB568123 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BDB568122 Supplier BD Horizon™ Supplier No. 568122
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

The HA58 monoclonal antibody specifically binds to CD54 which is also known as ICAM-1 (intracellular adhesion molecule-1).  CD54 is an 85-110 kDa type I transmembrane glycoprotein that belongs to the immunoglobulin supergene family. A soluble form of CD54 can also be found in biological fluids. CD54 is expressed on endothelial cells and both resting (weak) and activated (moderate) lymphocytes and monocytes. CD54 is a ligand for LFA-1 (CD11a/CD18) and Mac-1 (CD11b/CD18). The CD54 adhesion molecule plays roles in inflammatory and immune responses and neoplasia. This antibody (NA/LE format) blocks the mixed-lymphocyte reaction (MLR) and the purified format is suitable for staining acetone-fixed, frozen tissue sections.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD54 (ICAM-1)
Applications Flow Cytometry
Clone HA58
Conjugate RY586
Host Species Mouse
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.