Learn More
Description
Specifications
Specifications
| Assay Range | 23.44 pg/mL |
| Assay Sensitivity | 23.44-1500 pg/mL |
| Assay | Sandwich ELISA |
| Conjugate | HRP |
| Product Type | Antibody Pairs |
| Specificity | This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CD55/DAF. |
| Target Species | Human |
| For Use With (Application) | Sandwich ELISA |
| Gene Alias | CD55 antigen, CD55 molecule, decay accelerating factor for complement (Cromer blood group), CRdecay accelerating factor for complement (CD55, Cromer blood group system), CROMDAFcomplement decay-accelerating factor, decay accelerating factor for complement, TC |
| Gene ID (Entrez) | 1604 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
