Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD56 Mouse anti-Human, Brilliant Ultraviolet 737, Clone: NCAM16.2 (also known as NCAM 16), BD Horizon™

Catalog No. BDB612766 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612766 100 Tests
BDB612767 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB612766 Supplier BD Biosciences Supplier No. 612766
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The NCAM16.2 monoclonal antibody specifically binds to human CD56. It recognizes an extracellular immunoglobulin-like domain common to 120, 140, and 180 kDa forms of CD56, also known as the neural cell adhesion molecule (NCAM), NKH1 or MSK39. The CD56 antigen is expressed on approximately 10% to 25% of peripheral blood lymphocytes. It is present on essentially all resting and activated CD16+ natural killer (NK) lymphocytes and approximately 5% of CD3+ peripheral blood lymphocytes. CD3+ CD56+ T lymphocytes comprise a unique subset of cytotoxic T lymphocytes that mediates non-major histocompatibility complex (MHC)-restricted cytotoxicity. CD56 antigen density on NK lymphocytes increases upon cellular activation. The CD56 antigen is involved in neuronal homotypic cell adhesion and cell differentiation during embryogenesis. CD16+ CD56+ NK cells demonstrate reciprocal transfer of an activation state with dendritic cells.

The antibody was conjugated to BD Horizon™ BUV737 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye is a tandem fluorochrome of BD Horizon BUV395 with an Ex Max of 348-nm and an acceptor dye with an Em Max at 737-nm. BD Horizon Brilliant BUV737 can be excited by the ultraviolet laser (355 nm) and detected with a 740/35 filter. Due to the excitation of the acceptor dye by other laser lines, there may be significant spillover into channels detecting Alexa Fluor™ 700-like dyes (e.g., 712/20-nm filter).

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD56
Applications Flow Cytometry
Classification Monoclonal
Clone NCAM16.2 (also known as NCAM 16)
Conjugate Brilliant Ultraviolet 737
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Immunoaffinity-enriched adult human brain NCAM
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.