Learn More
Description
The antibody reacts with a 110 kD antigen present on the peripheral blood lymphocyte subset (primarily large granular lymphocytes) identified by the HNK-1 monoclonal antibody. It recognizes part of the Natural Killer cell population. CD57 is furthermore present on cells in neuroectodermal tissue.
Specifications
Specifications
| Antigen | CD57 |
| Applications | Immunohistochemistry (Frozen), Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 6-13-19-1 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide |
| Gene | B3GAT1 |
| Gene Accession No. | Q9P2W7 |
| Gene Alias | HNK-1; Leu-7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
