Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD5L Mouse, Rabbit anti-Human, Unconjugated, Abnova™

Catalog No. 89339487 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-487 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-487 Supplier Abnova Corporation Supplier No. H00000922PW2
Add to Cart
Add to Cart

Mouse, Rabbit Monoclonal/Polyclonal Antibody.

CD5L Monoclonal/Polyclonal antibody specifically detects CD5L in Human samples. It is validated for IP-WB.

Specifications

Antigen CD5L
Applications Immunoprecipitation, Western Blot
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description CD5 molecule-like
Gene CD5L
Gene Alias AIM, API6, PRO229, SP-ALPHA, Spalpha
Host Species Mouse, Rabbit
Quantity 1 Set
Primary or Secondary Primary
Gene ID (Entrez) 922
Includes Antibody pair for IP: mouse monoclonal anti-CD5L (300 μg); Antibody pair for WB: rabbit purified polyclonal anti-CD5L (50 μg)
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.