Learn More
Invitrogen™ CD61 (Integrin beta 3) Monoclonal Antibody (2C9.G3), NovaFluor™ Yellow 570, eBioscience™, Invitrogen™
Description
Specifications
Specifications
| Antigen | CD61 (Integrin beta 3) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 2C9.G3 |
| Concentration | 0.1 mg/mL |
| Conjugate | NovaFluor Yellow 570 |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | ITGB3 |
| Gene Accession No. | O54890 |
| Gene Alias | antigen CD61; BDPLT16; BDPLT2; beta3 integrin; CD61; EGK_08849; glycoprotein GPIIIa; glycoprotein IIIa; GP3A; GPIIIa; GT; INGRB3; integrin beta 3; integrin beta 3 (Cd61); integrin beta-3; integrin beta-3 subunit; integrin subunit beta 3; integrin, beta 3; integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61); ITGB3; platelet glycoprotein IIIa; platelet glycoprotein IIIa integrin subunit beta 3; platelet gpIIIa; platelet membrane glycoprotein IIIa |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.